Anti-MCP-2 (CCL8) Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P127BT-25UG 500-P127BT-50UG
10776-232EA 354.54 USD
10776-232 10776-234
Anti-MCP-2 (CCL8) Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Murine MCP-2 (CCL8). Anti­Murine MCP-2 (CCL8)­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: MCP-2 (CCL8)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR