Anti-TGF-alpha Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P16BT-25UG 500-P16BT-50UG
10775-064EA 339.8 USD
10775-064 10775-066 10775-068
Anti-TGF-alpha Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human TGF-α. Anti­Human TGF-α­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: TGF-alpha
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR