Anti-I-TAC (CXCL11) Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P132BT-25UG 500-P132BT-50UG
10776-292EA 339.8 USD
10776-292 10776-294 10776-296
Anti-I-TAC (CXCL11) Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human I-TAC (CXCL11). Anti­Human I-TAC (CXCL11)­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: I-TAC (CXCL11)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR