Anti-IL-17E Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P89BT-25UG 500-P89BT-50UG
10775-840EA 339.8 USD
10775-840 10775-842 10775-844
Anti-IL-17E Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human IL-17E. Anti­Human IL-17E­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: IL-17E
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR