Anti-Exodus-2 (CCL21) Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P109BT-25UG 500-P109BT-50UG
10776-004EA 368.12 USD
10776-004 10776-006 10776-008
Anti-Exodus-2 (CCL21) Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human Exodus-2 (CCL21). Anti­Human Exodus-2 (CCL21)­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: Exodus-2 (CCL21)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR