Anti-IFN-gamma Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P122BT-25UG 500-P122BT-50UG
10776-160EA 339.8 USD
10776-160 10776-162 10776-164
Anti-IFN-gamma Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Rat IFN-γ. Anti­Rat IFN-γ­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: IFN-gamma
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR