Anti-MIP-1alpha (CCL3) Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P77-50UG 500-P77-100UG 500-P77-1MG
10775-702EA 354.54 USD
10775-702 10775-704 10775-706
Anti-MIP-1alpha (CCL3) Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Rat MIP-1α (CCL3). Anti­Rat MIP-1α (CCL3)­specific antibody was purified by affinity chromatography employing an immobilized Rat MIP-1α matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: MIP-1alpha (CCL3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR