Anti-MCP-1 (CCL2) Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P76-50UG 500-P76-100UG 500-P76-1MG
10775-690EA 339.8 USD
10775-690 10775-692 10775-694
Anti-MCP-1 (CCL2) Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Rat MCP-1 (CCL2). Anti­Rat MCP-1 (CCL2)­specific antibody was purified by affinity chromatography employing an immobilized Rat MCP-1 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: MCP-1 (CCL2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR