Anti-MIP-2 (CXCL2) Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P130-50UG 500-P130-100UG 500-P130-1MG
10776-262EA 339.8 USD
10776-262 10776-264 10776-266
Anti-MIP-2 (CXCL2) Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Murine MIP-2 (CXCL2). Anti­Murine MIP-2 (CXCL2)­specific antibody was purified by affinity chromatography employing an immobilized Murine MIP-2 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: MIP-2 (CXCL2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR