Anti-Leptin Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P68-50UG 500-P68-100UG 500-P68-1MG
10775-594EA 339.8 USD
10775-594 10775-596 10775-598
Anti-Leptin Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Murine Leptin. Anti­Murine Leptin­specific antibody was purified by affinity chromatography employing an immobilized Murine Leptin matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: Leptin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR