Anti-IL-2 Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P111-50UG 500-P111-100UG 500-P111-1MG
10776-022EA 368.12 USD
10776-022 10776-024 10776-026
Anti-IL-2 Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Murine IL-2. Anti­Murine IL-2­specific antibody was purified by affinity chromatography employing an immobilized Murine IL-2 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IL-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR