Anti-IL-1beta Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P51-50UG 500-P51-100UG 500-P51-1MG
10775-422EA 339.8 USD
10775-422 10775-424 10775-426
Anti-IL-1beta Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Murine IL-1β. Anti­Murine IL-1β­specific antibody was purified by affinity chromatography employing an immobilized Murine IL-1β matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IL-1beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR