Anti-Wnt-3a Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P251-50UG 500-P251-100UG 500-P251-1MG
10777-266EA 339.8 USD
10777-266 10777-268 10777-270
Anti-Wnt-3a Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human Wnt-3a. Anti­Human Wnt-3a­specific antibody was purified by affinity chromatography employing an immobilized Human Wnt-3a matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: Wnt-3a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR