Anti-IP-10 (CXCL10) Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P93-50UG 500-P93-100UG 500-P93-1MG
10775-882EA 354.54 USD
10775-882 10775-884 10775-886
Anti-IP-10 (CXCL10) Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human IP-10 (CXCL10). Anti­Human IP-10 (CXCL10)­specific antibody was purified by affinity chromatography employing an immobilized Human IP-10 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IP-10 (CXCL10)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR