Anti-I-309 (CCL1) Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P110-50UG 500-P110-100UG 500-P110-1MG
10776-010EA 368.12 USD
10776-010 10776-012 10776-014
Anti-I-309 (CCL1) Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human I-309 (CCL1). Anti­Human I-309 (CCL1)­specific antibody was purified by affinity chromatography employing an immobilized Human I-309 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: I-309 (CCL1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR