Anti-GRO-gamma (CXCL3) Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P105-1MG 500-P105-50UG 500-P105-100UG
10771-328EA 2751.02 USD
10771-328 10775-998 10776-000
Anti-GRO-gamma (CXCL3) Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human GRO-γ (CXCL3). Anti­Human GRO-γ (CXCL3)­specific antibody was purified by affinity chromatography employing an immobilized Human GRO-γ matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: GRO-gamma (CXCL3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR