Anti-G-CSF Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P43-50UG 500-P43-100UG 500-P43-1MG
10775-314EA 368.12 USD
10775-314 10775-316 10775-318
Anti-G-CSF Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human G-CSF. Anti­Human G-CSF­specific antibody was purified by affinity chromatography employing an immobilized Human G-CSF matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: G-CSF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR