Anti-FGF-acidic Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P17-50UG 500-P17-100UG 500-P17-1MG
10775-070EA 339.8 USD
10775-070 10775-072 10775-074
Anti-FGF-acidic Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human FGF-acidic. Anti­Human FGF-acidic­specific antibody was purified by affinity chromatography employing an immobilized Human FGF-acidic matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: FGF-acidic
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR