Anti-BMP-7 Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P198-50UG 500-P198-100UG 500-P198-1MG
10776-818EA 339.8 USD
10776-818 10776-820 10776-822
Anti-BMP-7 Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human BMP-7. Anti­Human BMP-7­specific antibody was purified by affinity chromatography employing an immobilized Human BMP-7 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: BMP-7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR