Anti-IL-6 Goat Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P73GBT-25UG 500-P73GBT-50UG
10775-660EA 339.8 USD
10775-660 10775-662 10775-664
Anti-IL-6 Goat Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Rat IL-6. Anti­Rat IL-6­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: IL-6
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR