Anti-Resistin Goat Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P183GBT-25UG 500-P183GBT-50UG
10776-712EA 339.8 USD
10776-712 10776-714 10776-716
Anti-Resistin Goat Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Human Resistin. Anti­Human Resistin­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: Resistin
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR