Anti-IL-3beta Goat Polyclonal Antibody

Supplier: Peprotech
500-P177G-50UG 500-P177G-100UG 500-P177G-1MG
10776-646EA 339.8 USD
10776-646 10776-648 10776-650
Anti-IL-3beta Goat Polyclonal Antibody
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Rat IL-3β. Anti­Rat IL-3β­specific antibody was purified by affinity chromatography employing an immobilized Rat IL-3β matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IL-3beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR