Anti-TPO Goat Polyclonal Antibody

Supplier: Peprotech
500-P49G-50UG 500-P49G-100UG 500-P49G-1MG
10775-398EA 339.8 USD
10775-398 10775-400 10775-402
Anti-TPO Goat Polyclonal Antibody
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Human TPO. Anti­Human TPO­specific antibody was purified by affinity chromatography employing an immobilized Human TPO matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: TPO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR