Anti-sRANK Ligand Goat Polyclonal Antibody

Supplier: Peprotech
500-P133G-50UG 500-P133G-100UG 500-P133G-1MG
10776-310EA 339.8 USD
10776-310 10776-312 10776-314
Anti-sRANK Ligand Goat Polyclonal Antibody
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Human sRANK Ligand. Anti­Human sRANK Ligand­specific antibody was purified by affinity chromatography employing an immobilized Human sRANK Ligand matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: sRANK Ligand
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR