Anti-sCD40 Ligand Goat Polyclonal Antibody

Supplier: Peprotech
500-P142G-50UG 500-P142G-100UG 500-P142G-1MG
10776-366EA 354.54 USD
10776-366 10776-368 10776-370
Anti-sCD40 Ligand Goat Polyclonal Antibody
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Human sCD40 Ligand. Anti­Human sCD40 Ligand­specific antibody was purified by affinity chromatography employing an immobilized Human sCD40 Ligand matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: sCD40 Ligand
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR