Anti-EG-VEGF Goat Polyclonal Antibody

Supplier: Peprotech
500-P188G-50UG 500-P188G-100UG 500-P188G-1MG
10776-734EA 339.8 USD
10776-734 10776-736 10776-738
Anti-EG-VEGF Goat Polyclonal Antibody
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Human EG-VEGF. Anti­Human EG-VEGF­specific antibody was purified by affinity chromatography employing an immobilized Human EG-VEGF matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: EG-VEGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR