Stainless Steel Hooked Weight Sets
Supplier: United Scientific Supplies
WSST09 WSST10
470304-048EA 108.6 USD
470304-048 470304-050
Stainless Steel Hooked Weight Sets
Weights Calibration Weights
High Quality for the Classroom.
  • Stainless steel
  • Individually calibrated
Individually calibrated weights made of high quality stainless steel.
Both sets come with a sturdy plastic storage block. Set of 9 includes one each of 5 g, 10 g, 50 g, 100 g, and 500 g, plus two each of 20 g and 200 g weights.
Set of 10 includes all of the above, plus one each 1000 g weight.