Anti-MHC class II Monoclonal Antibody

Supplier: Bio X Cell
BE0306-1MG BE0306-100MG BE0306-25MG BE0306-50MG BE0306-5MG
103572-688EA 3920 USD
103572-688 103572-690 103572-692 103572-694 103572-696 76554-448 76555-008 76555-672 76556-328 76557-270
Anti-MHC class II Monoclonal Antibody
Antibodies
InVivoMAb anti-human/monkey MHC class II (HLA-DR), Clone: L243, Isotype: Mouse IgG2a, κ, Applications: in vitro blocking of MHC class II HLA-DR, HLA class II binding assay, in vitro MHC class II HLA-DR expressing cell negative selection, Western blot, Flow cytometry

Type: Primary
Antigen: MHC class II
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope: Mouse
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR