Accuris Mammalian Genotyping Kits
Supplier: Ward's
PR1300-MG-80 PR1300-MG-400
470305-348EA 149.9 USD
470305-348 470305-350
Accuris Mammalian Genotyping Kits
Educational Classroom Kits and Activities
This one-hour Mammalian Genotyping kit offers fast, efficient extraction and amplification of genomic DNA. The kit contains everything needed, just add sample and primers.
  • DNA extraction and amplification in an hour
  • Proprietary lysis buffer optimized for ear punches, tail snips, and other mammalian tissues
  • Single tube and no organic solvents or clean up procedures required
  • Enhanced Accuris Hot Start polymerase included
The Accuris Mammalian Genotyping Kit is quick and easy to use. Simply add sample to the proprietary lysis buffer and incubate for 5 to 10 minutes, then add the deactivation buffer and incubate for 10 minutes. The crude lysate can then be amplified using fast PCR Accuris Hot Start Taq Master Mix with red loading dye (included).
Delivery information: Upon receipt, immediately store at –20 °C. Avoid excessive freeze/thaw cycles. When stored as directed, this product will retain its activity for 12 months from date of receipt. Product can be stored at 4 °C for up to one month.
Caution: For research and educational purposes only. Not intended for therapeutic or diagnostic use.