Hydrocortisone ≥97%, white powder, Cell Culture Grade
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
0219456901
0219456905
0219456910
76204-372EA
65.1
USD
76204-372
76204-374
76204-376
Hydrocortisone ≥97%, white powder, Cell Culture Grade
Hydrocortisone
Hydrocortisone, a primary glucocorticoid secreted by the adrenal cortex. It has three times the anti-inflammatory potency of corticosterone but much lower Na+ retention potency. Hydrocortisone, known more formally as cortisone (INN, USAN, BAN), is a steroid hormone, produced by the zona fasciculata of the adrenal gland. It is released in response to stress and a low level of blood glucocorticoids.
- Grade : Cell Culture Reagent
- Purity : ≥97%
- Suitability : Suitable for cell culture
- Key Applications : Growth Factors, Hormones
- Application Areas : Molecular biology And Cell biology
- Product Type : Biochemicals
- Biochemical Category : Biochemiclas
- Optical Rotation : +160 ± 10 ° (c=1, methanol)
- UV/Visible Absorbance : 241 ± 3 nm (methanol)
- Presentation : White Powder
- Format Powder
- Loss on Drying : ≤1%
- Solubility : Soluble (mg/mL) at 25°C in: water 0.28; ethanol 15.0; methanol 6.2; acetone 9.3; chloroform 1.6; propylene glycol 12.7; ether ~ 0.35. Soluble in concentrated sulfuric acid with intense green fluorescence.
- Sterilization of Solutions: Solutions can be sterile filtered using a 0.2 µm membrane.
Storage: protect from light.
Formula:
C₂₁H₃₀O₅ MW: 362.47 g/mol Melting Pt: 205...222 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011654 CAS Number: 50-23-7 |
Specification Test Results
Identity Test | Passes |
FTIR | Conforms to Standard |
Melting Point | 205-222°C |
Loss on Drying |
|
UV/VIS: l max | 241 ± 3 nm (methanol) |
Optical Rotation (c=1, methanol) | +160 ± 10° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...