Hygromycin B (from Streptomyces hygroscopicus), >1000 u/mg ≥95%, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: (3aS,3'R,4S,4'S,5'R,6R,6'R,7S,7aS)-4-(((1R,2S,3R,5S,6R)-3-Amino-2,6-dihydroxy-5-(methylamino)cyclohexyl)oxy)-6'-((S)-1-amino-2-hydroxyethyl)-6-(hydroxymethyl)octahydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-2,2'-pyran]-3',4',5',7-tetraol

0219417050 0219417080 0219417091
76203-974EA 60.18 USD
76203-974 76203-976 76204-336
Hygromycin B (from Streptomyces hygroscopicus), >1000 u/mg ≥95%, white powder
Hygromycin B

Hygromycin B is an antibiotic substance isolated from Streptomyces hygroscopicus. Its mode of action is the inhibition of protein synthesis, by inducing the misreading of the m-RNA template in the prokaryote, (E. coli, at 100 mg/mL), lower eukaryotes (e.g., yeast, at 200 mg/mL) and higher eukaryotes (e.g., mammalian cells in culture). It selectively penetrates cells that have been rendered permeable by virus infection and combined with its potency to inhibit translation, it is an effective antiviral agent. Hygromycin B can be used as a dual-selection antibiotic when used in conjunction with G418 or Zeocin™ for cells transfected with two different vectors.


  • Purity : ~95%
  • Specific Activity : >1000 u/mg
  • Key Applications : In gene transfer experiments as a selection agent.
  • Application Areas : Cell Culture
  • Research Areas : Immunology, Cell Biology, Gene expression
  • Product Type : Biochemicals
  • Biochemical Category : Antibiotics
  • Chemical Class : Pharmaceuticals
  • Presentation : White Powder
  • pKa : 7.1
  • Detection Method : FTIR
  • Solubility : Freely soluble in water (50 mg/mL), methanol,buffer solution and ethanol. Practically insoluble in less polar solvents.

Formula: C₂₀H₃₇N₃O₁₃
MW: 527.5 g/mol
Melting Pt: 160...180 °C (lit.)
Storage Temperature: Refrigerator
MDL Number: MFCD06795479
CAS Number: 31282-04-9
UN: 3462
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d9a3680121619fec9bb9a96bdde48c2627f0e82&vid=8573127f1c687a3b3133c74b40c372663e6b1b3c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity >90%
Activity >1000 u/mg
FTIR Conforms to Reference


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR