Isoamyl alcohol ≥98.5%, clear, colourless liquid ACS
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
Isopentyl alcohol, iso-Pentanol, 3-Methyl-1-butanol, 3-Methylbutanol
0219405980
0219405990
76204-324EA
60.52
USD
76204-324
76204-326
Isoamyl alcohol ≥98.5%, clear, colourless liquid ACS
Isoamyl alcohol
- Grade : ACS
- Purity : ≥98.5%
- Key Applications : DNA purification; Conjugation
- Application Areas : Molecular biology
- Product Type : Biochemicals
- Auto Ignition : 350 °C (662 °F)(lit.)
- Vapor Density : 3.04 (air = 1) (lit.)
- Vapor Pressure : 2.4 mm Hg at 20 °C 2.37 mm Hg at 25 °C (lit.)
- Refractive Index : n20/D 1.406(lit.)
- Presentation : Clear, Colorless Liquid
- Format : Liquid
- Water Content : ≤0.5%
- NOTES : Titrable Acid <0.002 meq/g, Residue after Evaporation <0.003%, Acids and Esters (as amyl acetate) <0.2%, Carbonyl (as HCHO) <0.1%
- Solubility : Miscible with alcohol, ether, benzene, chloroform, petroleum ether, glacial acetic acid, oils.
Formula:
(CH₃)₂CHCH₂CH₂OH MW: 88.15 g/mol Boiling Pt: 130 (lit.) Melting Pt: -117 (lit.) Density: ∼0.81 g/mL Flash Pt: 45.5 °C (109.4 °F) (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00002934 CAS Number: 123-51-3 UN: 1105 ADR: 3 III Merck Index: 13,05214 |
|
Specification Test Results
Identity Test | Passes |
Purity |
|
Water |
|
Titrable Acid |
|
Residue after Evaporation |
|
Acids and Esters (as amyl acetate) |
|
Carbonyl (as HCHO) |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...