1-Butanol ≥99% for molecular biology

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: Propylcarbinol, Methylolpropane, Butan-1-ol, Butyl alcohol, n-Butyl alcohol, Butyl hydroxide, 1-Butyl alcohol, Butyl Alcohol, 1-Hydroxybutane, 1-Hydroxy butane, n-Butan-1-ol, Propyl carbinol, n-Butanol, n-Butyl Alcohol

0219400125 0219400191
76204-070EA 55.88 USD
76204-070 76204-292
1-Butanol ≥99% for molecular biology
1-Butanol

  • Grade Molecular biology reagent
  • Purity ≥99%
  • Key Applications Removal of ethidium bromide from DNA
  • Application Areas Molecular biology
  • Product Type Biochemicals
  • Auto Ignition 340 °C(Lit.)
  • Vapor Density 2.6 (air = 1)(Lit.)
  • Vapor Pressure 0.658 hPa at 20 °C(Lit.)
  • Refractive Index n20/D 1.399(lit.)
  • Presentation Liquid
  • Format Liquid
  • Detection Method FTIR
  • NOTES Japan Fire Code:4.2


Storage: under nitrogen.

Formula: H₃C(CH₂)₂CH₂OH
MW: 74.12 g/mol
Boiling Pt: 117.6 (lit.)
Melting Pt: -89 (lit.)
Density: 0.81 g/cm³ (21 °C) (lit.)
Flash Pt: 35 °C (95 °F) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00002964
CAS Number: 71-36-3
UN: 1120
Merck Index: 13,01539
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3bba269ef2db76a5a7efba22e720e9a25c7beafb&vid=1fbbac0be4a7398a251e9bf4d7de1abb334cecea&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity >99%
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR