Trimethylgallium ≥99.9999%, PURATREM, Electronic Grade

Supplier: STREM CHEMICALS INC MS
Danger

98-2000-50G 98-2000-100G
100203-658EA 6362.15 USD
100203-658 100203-656
Trimethylgallium ≥99.9999%, PURATREM, Electronic Grade
Trimethylgallium

Metal Alkyls, Volatile Precursors for CVD

Boiling Pt: 55.7 °C
Melting Pt: –15.8 °C
Density: 1.151
Flash Pt: –1 °F
MDL Number: MFCD00014841
CAS Number: 1445-79-0
UN: 3394
ADR: 4.2,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR