Cyclopentadienylindium(I) ≥99.99% (In basis), PURATREM, Electronic Grade

Supplier: STREM CHEMICALS INC MS
Danger

97-3425-5G 97-3425-1G
100203-642EA 2427.38 USD
100203-642 100200-358
Cyclopentadienylindium(I) ≥99.99% (In basis), PURATREM, Electronic Grade
Cyclopentadienylindium(I)

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Boiling Pt: 50 °C (0.01 mmHg) (subl.)
MDL Number: MFCD00151371
CAS Number: 34822-89-4
UN: 1325
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR