Hafnium dichloride oxide octahydrate ≥98.5% (Hf basis) (1.5% Zr)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Hafnium(IV) dichloride oxide octahydrate

93-7207-10G 93-7207-50G
100205-746EA 97.66 USD
100205-746 100203-580
Hafnium dichloride oxide octahydrate ≥98.5% (Hf basis) (1.5% Zr)
Hafnium dichloride oxide octahydrate
Formula: Cl₂HfO·8H₂O
MDL Number: MFCD00150417
CAS Number: 14456-34-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR