Samarium(III) oxalate hydrate ≥99.9% (Sm basis)

Supplier: STREM CHEMICALS INC MS

93-6209-10G 93-6209-50G
100196-792EA 77.85 USD
100196-792 100198-428
Samarium(III) oxalate hydrate ≥99.9% (Sm basis)
Samarium(III) oxalate hydrate
Formula: C₆O₁₂Sm₂.xH₂O
MDL Number: MFCD00150491
CAS Number: 14175-03-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR