Tin(IV) phthalocyanine dichloride

Supplier: STREM CHEMICALS INC MS

93-5012-5G 93-5012-1G
100207-094EA 242.03 USD
100207-094 100202-172
Tin(IV) phthalocyanine dichloride
Tin (IV) phthalocyanine dichloride
Formula: C₃₂H₁₆Cl₂N₈Sn
MDL Number: MFCD00054210
CAS Number: 18253-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR