Dibutyltin diacetate ≥96%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Di-n-butyltin diacetate
93-5007-100G
93-5007-500G
100209-138EA
92
USD
100209-138
100200-770
Dibutyltin diacetate ≥96%
Dibutyltin diacetate
Formula:
C₁₂H₂₄O₄Sn Melting Pt: 10 °C Density: 1.32 Flash Pt: >230 °F |
MDL Number:
MFCD00008697 CAS Number: 1067-33-0 UN: 2788 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...