Palladium(II) cyanide ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

93-4605-1G 93-4605-5G
100207-068EA 377.91 USD
100207-068 100207-070
Palladium(II) cyanide ≥98%
Palladium (II) cyanide
Formula: C₂N₂Pd
MDL Number: MFCD00016282
CAS Number: 2035-66-7
UN: 3288
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR