Molybdenum carbide ≥99.5% (Mo basis)
Supplier: STREM CHEMICALS INC MS
93-4243-50G
93-4243-250G
100202-132EA
84.92
USD
100202-132
100200-342
Molybdenum carbide ≥99.5% (Mo basis)
Molybdenum carbide
Formula:
CMo₂ Melting Pt: 2687 °C Density: 9,18 |
MDL Number:
MFCD00014218 CAS Number: 12069-89-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...