Tetrakis(trifluoro-2,4-pentanedionato)zirconium(IV) ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Zirconium(IV) trifluoroacetylacetonate, Zirconium(IV) Trifluoroacetylacetonate, Trifluoroacetylacetono Zirconium(IV) Salt
93-4026-25G
93-4026-5G
93-4026-1G
100205-564EA
512.37
USD
100205-564
100205-566
100204-324
Tetrakis(trifluoro-2,4-pentanedionato)zirconium(IV) ≥99%
Tetrakis(trifluoro-2,4-pentanedionato)zirconium(IV)
Metal Beta-diketonates, Volatile Precursors for CVD
Formula:
C₂₀H₁₆F₁₂O₈Zr Boiling Pt: 235 °C/ 130 °C (0.05 mmHg) (dec./ subl.) Melting Pt: 125…128 °C |
MDL Number:
MFCD00015317 CAS Number: 17499-68-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...