Nickel(II) trifluoroacetylacetonate dihydrate ≥98%

Supplier: STREM CHEMICALS INC MS

93-2843-25G 93-2843-5G
100204-694EA 982.28 USD
100204-694 100210-328
Nickel(II) trifluoroacetylacetonate dihydrate ≥98%
Nickel(II) trifluoroacetylacetonate dihydrate
MDL Number: MFCD00150656
CAS Number: 14324-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR