Nickel hydroxyacetate

Supplier: STREM CHEMICALS INC MS

Synonyms: Nickel(II) hydroxyacetate

93-2826-5G 93-2826-25G
100204-280EA 107.57 USD
100204-280 100199-046
Nickel hydroxyacetate
Nickel hydroxyacetate
Formula: C₄H₆NiO₆
MDL Number: MFCD00070414
CAS Number: 41587-84-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR