Nickel acetate tetrahydrate ≥98%

Supplier: STREM CHEMICALS INC MS

93-2803-1000G 93-2803-250G
100211-242EA 328.37 USD
100211-242 100197-790
Nickel acetate tetrahydrate ≥98%
Nickel acetate tetrahydrate
Formula: C₄H₆NiO₄·4H₂O
Melting Pt: Dec.
Density: 1.744
MDL Number: MFCD00066973
CAS Number: 6018-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR