Tris(ethylenediamine)chromium(III) chloride hemiheptahydrate ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-2444-25G
93-2444-5G
100197-202EA
369.42
USD
100197-202
100207-800
Tris(ethylenediamine)chromium(III) chloride hemiheptahydrate ≥98%
Tris(ethylenediamine)chromium(III) chloride hemiheptahydrate
Formula:
C₆H₂₄Cl₃CrN₆.3,5H₂O |
MDL Number:
MFCD00150160 CAS Number: 16165-32-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...