Calcium methoxide ≥90% (balance methanol)

Supplier: STREM CHEMICALS INC MS
Danger

93-2062-25G 93-2062-100G
100210-246EA 461.42 USD
100210-246 100199-000
Calcium methoxide ≥90% (balance methanol)
Calcium methoxide

Alkoxide

MDL Number: MFCD00058816
CAS Number: 2556-53-8
UN: 1325
ADR: 4,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR