Sodium metatitanate ≥95%

Supplier: STREM CHEMICALS INC MS

Synonyms: Sodium titanium oxide, Sodium titanate

93-1197-500G 93-1197-100G
100203-366EA 443.02 USD
100203-366 100205-294
Sodium metatitanate ≥95%
Sodium metatitanate
Formula: Na₂Ti₃O₇
Melting Pt: 1128 °C
Density: 3.35…3.50
MDL Number: MFCD00014243
CAS Number: 12034-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR