tert-Butyllithium 16% (1-2M) in pentane

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: t-Butyllithium

93-0302-1MOL 93-0302-250MMOL
100206-872EA 389.23 USD
100206-872 100206-870
tert-Butyllithium 16% (1-2M) in pentane
tert-Butyllithium
Formula: C₄H₉Li
MDL Number: MFCD00008795
CAS Number: 594-19-4
UN: 3394
ADR: 4.2,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR