Hexaphenyldilead ≥98%

Supplier: STREM CHEMICALS INC MS

82-0580-5G 82-0580-1G
100206-000EA 295.82 USD
100206-000 100201-774
Hexaphenyldilead ≥98%
Hexaphenyldilead
Formula: C₃₆H₃₀Pb₂
Melting Pt: 170 °C (dec.)
MDL Number: MFCD00014066
CAS Number: 3124-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR